Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh3
Entrez Gene ID
12560
Gene Name
cadherin 3
Gene Aliases
AI385538, Cadp, Cdhp, P-cadherin, Pcad
Chromosome Location
Chr.8: 106510852 - 106556911 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037809.5 NP_001032898.1 15 - 16 2364 71
NM_007665.3 NP_031691.1 15 - 16 2361 71
GenBank mRNA AK031265.1 - 15 - 16 2304 71
AK045041.1 - 15 - 16 2324 71
AK164577.1 - 15 - 16 2334 71
BC052189.1 - 13 - 14 2052 71
BC098459.1 - 15 - 16 2270 71
X06340.1 - 15 - 16 71

View Full Product Details