The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_148940.3 | NP_683742.2 | 2 - 3 | 515 | 79 |
GenBank mRNA | AB162857.1 | - | 2 - 3 | 501 | 79 |
AK006612.1 | - | 2 - 3 | 515 | 79 |