Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Commd7
Entrez Gene ID
99311
Gene Name
COMM domain containing 7
Gene Aliases
2310010I22Rik, AW060956, mU3
Chromosome Location
Chr.2: 153616930 - 153632794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195390.1 NP_001182319.1 1 - 2 183 58
GenBank mRNA AK169037.1 - 1 - 2 130 58
AK177311.1 - 1 - 2 32 58

View Full Product Details