Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Has3
Entrez Gene ID
15118
Gene Name
hyaluronan synthase 3
Gene Aliases
-
Chromosome Location
Chr.8: 106870242 - 106882902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK028582.1 - 1 - 2 121 99

View Full Product Details