Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbs2
Entrez Gene ID
67378
Gene Name
Bardet-Biedl syndrome 2 (human)
Gene Aliases
2410125H22Rik, AI447581
Chromosome Location
Chr.8: 94067954 - 94099370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026116.2 NP_080392.1 14 - 15 2250 76
GenBank mRNA AF342737.1 - 14 - 15 2250 76
AK010779.1 - 14 - 15 1863 76
BC057184.1 - 14 - 15 76

View Full Product Details