Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gnao1
Entrez Gene ID
14681
Gene Name
guanine nucleotide binding protein, alpha O
Gene Aliases
AW050213, Galphao, Gnao, alphaO
Chromosome Location
Chr.8: 93810561 - 93969388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113384.1 NP_001106855.1 2 - 3 653 113
NM_010308.3 NP_034438.1 2 - 3 653 113
GenBank mRNA AK035907.1 - 2 - 3 654 113
AK047197.1 - 2 - 3 657 113
BC051989.1 - 2 - 3 480 113
M36777.1 - 2 - 3 176 113
M36778.1 - 2 - 3 178 113

View Full Product Details