Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nts
Entrez Gene ID
67405
Gene Name
neurotensin
Gene Aliases
5033428E16Rik, NMN-125, NN, NT, NT/N, NTS1
Chromosome Location
Chr.10: 102481756 - 102490418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024435.2 NP_077755.1 1 - 2 161 69
GenBank mRNA AF304160.1 - 1 - 2 164 69
AK017202.1 - 1 - 2 178 69
AK017212.1 - 1 - 2 177 69
BC043024.1 - 1 - 2 161 69

View Full Product Details