The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024435.2 | NP_077755.1 | 3 - 4 | 453 | 150 |
GenBank mRNA | AK017202.1 | - | 3 - 4 | 471 | 150 |
AK017212.1 | - | 3 - 4 | 469 | 150 | |
BC043024.1 | - | 3 - 4 | 453 | 150 |