Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dhx8
Entrez Gene ID
217207
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 8
Gene Aliases
Ddx8, mDEAH6, mKIAA4096
Chromosome Location
Chr.11: 101732956 - 101767358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144831.2 NP_659080.2 14 - 15 2263 79
GenBank mRNA AK078552.1 - 14 - 15 2202 79
AK142226.1 - 14 - 15 2197 79
AK160848.1 - - 204 79
AK172600.1 - 14 - 15 2211 79
AK220485.1 - 14 - 15 2240 79
BC065064.1 - 14 - 15 2192 79
BC132445.1 - 14 - 15 2247 79

View Full Product Details