Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhx8
Entrez Gene ID
217207
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 8
Gene Aliases
Ddx8, mDEAH6, mKIAA4096
Chromosome Location
Chr.11: 101732956 - 101767358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144831.2 NP_659080.2 4 - 5 474 61
GenBank mRNA AK078552.1 - 4 - 5 413 61
AK142226.1 - 4 - 5 408 61
AK153829.1 - 4 - 5 475 61
AK172600.1 - 4 - 5 414 61
AK220485.1 - 4 - 5 433 61
BB654249.1 - 4 - 5 475 61
BC065064.1 - 4 - 5 403 61
BC132445.1 - 4 - 5 458 61

View Full Product Details