Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ncoa6
- Entrez Gene ID
-
56406
- Gene Name
- nuclear receptor coactivator 6
- Gene Aliases
-
AIB3, ASC-2, ASC2, NRC, Ncoa7, PRIP, RAP250, mKIAA0181
- Chromosome Location
-
Chr.2: 155390656 - 155473894 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK144338.1 |
- |
2 - 3 |
232 |
83 |
View Full Product Details