The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164677.1 | NP_001158149.1 | 5 - 6 | 762 | 77 |
NM_001164678.1 | NP_001158150.1 | 5 - 6 | 762 | 77 | |
NM_011052.2 | NP_035182.2 | 5 - 6 | 762 | 77 | |
GenBank mRNA | AF119955.1 | - | 5 - 6 | 723 | 77 |
AJ005073.1 | - | 5 - 6 | 610 | 77 | |
AK029827.1 | - | 5 - 6 | 729 | 77 | |
AK033943.1 | - | 5 - 6 | 751 | 77 | |
AK129340.1 | - | 5 - 6 | 732 | 77 | |
AK145893.1 | - | 5 - 6 | 750 | 77 | |
AK147736.1 | - | 5 - 6 | 735 | 77 | |
AK167574.1 | - | 5 - 6 | 720 | 77 | |
AK169704.1 | - | 5 - 6 | 763 | 77 | |
AK207830.1 | - | 2 - 3 | 167 | 77 | |
BC051123.1 | - | 5 - 6 | 613 | 77 |