The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081043.1 | NP_001074512.1 | 2 - 3 | 254 | 77 |
GenBank mRNA | CF538443.1 | - | 2 - 3 | 215 | 77 |