Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsf3
Entrez Gene ID
257633
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
BB101783
Chromosome Location
Chr.8: 122775493 - 122817882 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144932.3 NP_659181.2 7 - 8 1475 68
GenBank mRNA AK144352.1 - 8 - 9 1607 68
BC021611.1 - 3 - 4 348 68
BC022709.1 - 6 - 7 815 68

View Full Product Details