Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam20a
Entrez Gene ID
208659
Gene Name
family with sequence similarity 20, member A
Gene Aliases
AI606893
Chromosome Location
Chr.11: 109669492 - 109722510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153782.1 NP_722477.1 7 - 8 1651 79
GenBank mRNA AK142932.1 - 7 - 8 1676 79
BC011274.1 - 7 - 8 1555 79
BC029169.1 - 7 - 8 1651 79

View Full Product Details