The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010303.3 | NP_034433.3 | 1 - 2 | 467 | 63 |
GenBank mRNA | AK011851.1 | - | 1 - 2 | 430 | 63 |
AK149766.1 | - | 1 - 2 | 361 | 63 | |
BC057665.1 | - | 1 - 2 | 321 | 63 | |
M63660.1 | - | 1 - 2 | 415 | 63 |