Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tm4sf5
Entrez Gene ID
75604
Gene Name
transmembrane 4 superfamily member 5
Gene Aliases
2010003F10Rik, AI323987
Chromosome Location
Chr.11: 70505274 - 70511183 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029360.3 NP_083636.2 4 - 5 632 58
GenBank mRNA AI323987.1 - 1 - 2 58
BB798656.1 - 1 - 2 51 58
BC010782.1 - 4 - 5 592 58

View Full Product Details