The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020510.2 | NP_065256.1 | 1 - 1 | 2722 | 80 |
GenBank mRNA | AK048420.1 | - | 1 - 1 | 80 | |
AK135363.1 | - | 1 - 1 | 2686 | 80 | |
BC049774.1 | - | 1 - 1 | 2539 | 80 |