The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013640.3 | NP_038668.2 | 7 - 8 | 1097 | 79 |
GenBank mRNA | AK144096.1 | - | 7 - 8 | 820 | 79 |
BC004730.1 | - | 7 - 8 | 823 | 79 | |
BY686068.1 | - | 3 - 4 | 238 | 79 | |
CT010274.1 | - | 7 - 8 | 715 | 79 | |
D85561.1 | - | 7 - 8 | 792 | 79 | |
U77784.1 | - | 7 - 8 | 784 | 79 | |
Y10875.1 | - | 7 - 8 | 815 | 79 |