Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dpep2
Entrez Gene ID
319446
Gene Name
dipeptidase 2
Gene Aliases
F630103D06Rik, MBD-2
Chromosome Location
Chr.8: 105984945 - 106006348 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301204.1 NP_001288133.1 2 - 3 439 112
NM_176913.4 NP_795887.2 2 - 3 554 112
GenBank mRNA AF488552.1 - 2 - 3 554 112
AK089244.1 - 2 - 3 425 112
AK155308.1 - 2 - 3 112
AK155417.1 - 2 - 3 394 112
AK170094.1 - 2 - 3 391 112
AK170970.1 - 2 - 3 391 112
AK171044.1 - 2 - 3 391 112
AK171122.1 - 2 - 3 391 112
AK171196.1 - 2 - 3 391 112
AK171268.1 - 2 - 3 391 112
AK171503.1 - 2 - 3 391 112

View Full Product Details