The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176913.4 | NP_795887.2 | 4 - 5 | 829 | 74 |
GenBank mRNA | AF488552.1 | - | 4 - 5 | 829 | 74 |