Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsnaxip1
Entrez Gene ID
72236
Gene Name
translin-associated factor X (Tsnax) interacting protein 1
Gene Aliases
1700016K08Rik, TXI1
Chromosome Location
Chr.8: 105827711 - 105844676 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024445.4 NP_077765.1 15 - 16 1920 67
GenBank mRNA AF343750.1 - 15 - 16 1892 67
AK006041.1 - 15 - 16 1921 67

View Full Product Details