The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027468.1 | NP_081744.1 | 6 - 7 | 810 | 75 |
GenBank mRNA | BC047389.1 | - | 5 - 6 | 796 | 75 |
BC055073.1 | - | 6 - 7 | 815 | 75 | |
BC088735.1 | - | 6 - 7 | 815 | 75 | |
BC100404.1 | - | 6 - 7 | 804 | 75 |