The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141924.1 | NP_001135396.1 | 3 - 4 | 849 | 117 |
GenBank mRNA | AK134633.1 | - | 3 - 4 | 849 | 117 |