Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbfb
Entrez Gene ID
12400
Gene Name
core binding factor beta
Gene Aliases
AI893578, PEA2, PEBP2b, Pebp2, Pebpb2
Chromosome Location
Chr.8: 105170674 - 105217989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161456.1 NP_001154928.1 3 - 4 585 60
NM_001161458.1 NP_001154930.1 4 - 5 702 60
NM_022309.4 NP_071704.3 4 - 5 702 60
GenBank mRNA AK089094.1 - 4 - 5 703 60
AK134449.1 - 3 - 4 270 60
AK198845.1 - - 152 60
BC006763.1 - 4 - 5 470 60
BC026749.1 - 4 - 5 512 60
BC040752.1 - 4 - 5 501 60
D14571.1 - 4 - 5 632 60
D14572.1 - 4 - 5 632 60
L03279.1 - 4 - 5 481 60
L03305.1 - 3 - 4 323 60
L03306.1 - - 400 60

View Full Product Details