Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc9a5
Entrez Gene ID
277973
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 5
Gene Aliases
Gm696
Chromosome Location
Chr.8: 105348104 - 105369881 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081332.1 NP_001074801.1 6 - 7 2259 58
GenBank mRNA BC151197.1 - 6 - 7 1700 58

View Full Product Details