The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175160.3 | NP_780369.1 | 3 - 4 | 179 | 73 |
GenBank mRNA | AK038593.1 | - | 3 - 4 | 181 | 73 |