Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbfa2t2
Entrez Gene ID
12396
Gene Name
core-binding factor, runt domain, alpha subunit 2, translocated to, 2 (human)
Gene Aliases
A430091M07, C330013D05Rik, Cbfa2t2h, MTGR1
Chromosome Location
Chr.2: 154436463 - 154539356 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285446.1 NP_001272375.1 10 - 11 1608 71
NM_009823.1 NP_033953.1 10 - 11 1609 71
NM_172860.2 NP_766448.1 10 - 11 1612 71
GenBank mRNA AK040403.1 - 10 - 11 1601 71
AK049206.1 - 10 - 11 1570 71
AK153602.1 - 10 - 11 1514 71
BC064679.1 - 10 - 11 1578 71
BC138410.1 - 10 - 11 1545 71
BC145266.1 - 10 - 11 1541 71
BC145679.1 - 10 - 11 1545 71
DQ213025.1 - 10 - 11 1585 71

View Full Product Details