Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gan
Entrez Gene ID
209239
Gene Name
giant axonal neuropathy
Gene Aliases
A330045G18, gigaxonin
Chromosome Location
Chr.8: 117158135 - 117205186 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081151.1 NP_001074620.1 8 - 9 1377 69
GenBank mRNA AK039455.1 - 8 - 9 1376 69
BC112432.1 - 8 - 9 1376 69

View Full Product Details