Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfs1
Entrez Gene ID
18041
Gene Name
nitrogen fixation gene 1 (S. cerevisiae)
Gene Aliases
AA987187, m-Nfs1, m-Nfsl
Chromosome Location
Chr.2: 156123637 - 156144186 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010911.2 NP_035041.2 6 - 7 704 93
GenBank mRNA AK005724.1 - 6 - 7 693 93
AK075575.1 - 6 - 7 702 93
AK132838.1 - 6 - 7 705 93
AK159828.1 - 6 - 7 693 93
AK183689.1 - 2 - 3 110 93
AK206231.1 - 2 - 3 110 93
AK206381.1 - 2 - 3 110 93
AK206572.1 - 2 - 3 110 93
AK207148.1 - 2 - 3 110 93
AK207847.1 - 2 - 3 110 93
AK207895.1 - 2 - 3 110 93
AK211411.1 - 2 - 3 110 93
AK211628.1 - 2 - 3 110 93
AK212905.1 - 2 - 3 110 93
AK216163.1 - 1 - 2 110 93
AK220082.1 - 1 - 2 110 93
BC010586.1 - 6 - 7 93

View Full Product Details