Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cherp
Entrez Gene ID
27967
Gene Name
calcium homeostasis endoplasmic reticulum protein
Gene Aliases
5730408I11Rik, D8Wsu96e, DAN16, Scaf6
Chromosome Location
Chr.8: 72460483 - 72476675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138585.3 NP_613051.3 10 - 11 1920 59
GenBank mRNA AF536541.1 - 10 - 11 1822 59
AK049765.1 - 10 - 11 1831 59
AK144328.1 - 10 - 11 1843 59
AK171907.1 - 3 - 4 896 59
AK187667.1 - 1 - 2 35 59
AK211710.1 - 1 - 2 35 59
BC032191.1 - 10 - 11 1838 59

View Full Product Details