The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016776.2 | NP_058056.2 | 3 - 4 | 377 | 114 |
GenBank mRNA | AK083908.1 | - | 3 - 4 | 401 | 114 |
AK143469.1 | - | 3 - 4 | 399 | 114 | |
AK143487.1 | - | 3 - 4 | 410 | 114 | |
AK154016.1 | - | 3 - 4 | 399 | 114 | |
AK155074.1 | - | 3 - 4 | 388 | 114 | |
AK166533.1 | - | 3 - 4 | 401 | 114 | |
AK208884.1 | - | 3 - 4 | 281 | 114 | |
BC015070.1 | - | 3 - 4 | 378 | 114 | |
BC048858.1 | - | 3 - 4 | 377 | 114 | |
BC052889.1 | - | 3 - 4 | 398 | 114 | |
BC060167.1 | - | 3 - 4 | 383 | 114 | |
U63648.1 | - | 3 - 4 | 386 | 114 |