Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cirh1a
Entrez Gene ID
21771
Gene Name
cirrhosis, autosomal recessive 1A (human)
Gene Aliases
Cirhin, Naic, Teg-292, Tex292
Chromosome Location
Chr.8: 106893636 - 106923094 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011574.2 NP_035704.2 3 - 4 428 86
GenBank mRNA AK144455.1 - 3 - 4 413 86
AK146263.1 - 3 - 4 404 86
AK150862.1 - 3 - 4 403 86
AK168470.1 - 3 - 4 378 86
AK173318.1 - 2 - 3 369 86
BC068113.1 - 3 - 4 417 86
CN718589.1 - 3 - 4 428 86

View Full Product Details