Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cirh1a
Entrez Gene ID
21771
Gene Name
cirrhosis, autosomal recessive 1A (human)
Gene Aliases
Cirhin, Naic, Teg-292, Tex292
Chromosome Location
Chr.8: 106893636 - 106923094 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011574.2 NP_035704.2 5 - 6 601 88
GenBank mRNA AK144455.1 - 5 - 6 586 88
AK146263.1 - 5 - 6 577 88
AK150862.1 - 5 - 6 576 88
AK168470.1 - 5 - 6 551 88
AK173318.1 - 4 - 5 542 88
AK178383.1 - 1 - 2 50 88
AK178714.1 - 1 - 2 50 88
AK180185.1 - 1 - 2 50 88
AK187526.1 - 1 - 2 88
BC027399.1 - 1 - 2 100 88
BC068113.1 - 5 - 6 590 88
CN718589.1 - 5 - 6 601 88

View Full Product Details