Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdr2l
Entrez Gene ID
237988
Gene Name
cerebellar degeneration-related protein 2-like
Gene Aliases
D030068L24Rik, Gm21
Chromosome Location
Chr.11: 115381916 - 115396132 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080929.1 NP_001074398.1 1 - 2 486 72
GenBank mRNA BC144823.1 - 1 - 2 356 72
BC150668.1 - 1 - 2 356 72

View Full Product Details