The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080929.1 | NP_001074398.1 | 3 - 4 | 747 | 68 |
GenBank mRNA | BC144823.1 | - | 3 - 4 | 617 | 68 |
BC150668.1 | - | 3 - 4 | 617 | 68 |