Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdr2l
Entrez Gene ID
237988
Gene Name
cerebellar degeneration-related protein 2-like
Gene Aliases
D030068L24Rik, Gm21
Chromosome Location
Chr.11: 115381916 - 115396132 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC144823.1 - 4 - 5 785 67

View Full Product Details