Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Recql5
Entrez Gene ID
170472
Gene Name
RecQ protein-like 5
Gene Aliases
RecQ5, Recq5b, Recql5b
Chromosome Location
Chr.11: 115892595 - 115933531 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130454.2 NP_569721.1 16 - 17 3127 72
GenBank mRNA AB059999.1 - 16 - 17 2641 72
AK049269.1 - 11 - 12 1527 72
AK050965.1 - 14 - 15 1856 72
BC019613.1 - 2 - 3 913 72
BC023769.1 - 15 - 16 2419 72
BC053086.1 - 16 - 17 3380 72
BC059000.1 - 16 - 17 3100 72
BC060070.1 - 16 - 17 3380 72
BC066176.1 - 16 - 17 2850 72

View Full Product Details