Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Qrich2
Entrez Gene ID
217341
Gene Name
glutamine rich 2
Gene Aliases
Gm66
Chromosome Location
Chr.11: 116441323 - 116466269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033267.2 NP_001028439.1 8 - 9 1205 67
GenBank mRNA AK131945.1 - 8 - 9 1208 67
BC150976.1 - 8 - 9 1058 67

View Full Product Details