Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim47
Entrez Gene ID
217333
Gene Name
tripartite motif-containing 47
Gene Aliases
2210023F24Rik
Chromosome Location
Chr.11: 116105750 - 116110235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205081.1 NP_001192010.1 3 - 4 1027 57
NM_172570.5 NP_766158.3 3 - 4 1027 57
GenBank mRNA AK031572.1 - 3 - 4 1002 57
AK075838.1 - 3 - 4 456 57
BC023393.1 - 1 - 2 202 57
BC024468.1 - 1 - 2 71 57
BC043714.1 - 2 - 3 339 57
BC055038.1 - 1 - 2 327 57
BC064728.1 - 3 - 4 421 57
BC080731.1 - 3 - 4 423 57
BC100317.1 - 3 - 4 1011 57
BC132148.1 - 3 - 4 808 57
BC137953.1 - 3 - 4 808 57
BC145059.1 - 3 - 4 806 57

View Full Product Details