Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim47
Entrez Gene ID
217333
Gene Name
tripartite motif-containing 47
Gene Aliases
2210023F24Rik
Chromosome Location
Chr.11: 116105750 - 116110235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205081.1 NP_001192010.1 5 - 6 1304 131
NM_172570.5 NP_766158.3 5 - 6 1301 131
GenBank mRNA AK031572.1 - 5 - 6 1276 131
AK075838.1 - 5 - 6 730 131
BC023393.1 - 3 - 4 476 131
BC024468.1 - 3 - 4 345 131
BC043714.1 - 4 - 5 616 131
BC055038.1 - 3 - 4 601 131
BC064728.1 - 5 - 6 698 131
BC080731.1 - 5 - 6 700 131
BC100317.1 - 5 - 6 1285 131
BC132148.1 - 5 - 6 1082 131
BC137953.1 - 5 - 6 1082 131
BC145059.1 - 5 - 6 1083 131

View Full Product Details