Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Urb2
Entrez Gene ID
382038
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Gene Aliases
C77575, mKIAA0133
Chromosome Location
Chr.8: 124021442 - 124048505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029876.1 NP_001025047.1 6 - 7 4145 111
GenBank mRNA AK122209.1 - 4 - 5 3782 111
AK133906.1 - 7 - 8 4446 111
BC096020.1 - 4 - 5 2528 111
BC151125.1 - 6 - 7 4224 111

View Full Product Details