Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hgs
Entrez Gene ID
15239
Gene Name
HGF-regulated tyrosine kinase substrate
Gene Aliases
Hgr, Hrs, ZFYVE8
Chromosome Location
Chr.11: 120467635 - 120483984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159328.1 NP_001152800.1 19 - 20 2110 69
NM_008244.3 NP_032270.3 19 - 20 2107 69
GenBank mRNA AK046299.1 - 19 - 20 2068 69
AK145031.1 - 19 - 20 2030 69
AK166441.1 - 19 - 20 2081 69
BC003239.1 - 19 - 20 2051 69
D50050.1 - 19 - 20 2057 69

View Full Product Details