Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nploc4
Entrez Gene ID
217365
Gene Name
nuclear protein localization 4 homolog (S. cerevisiae)
Gene Aliases
AK129375, Npl4, mKIAA1499
Chromosome Location
Chr.11: 120379798 - 120437700 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195023.1 NP_001181952.1 13 - 14 1545 110
GenBank mRNA AK129375.1 - 13 - 14 1520 110
AK141998.1 - 13 - 14 1544 110
AK149823.1 - 13 - 14 1517 110
AK149912.1 - 13 - 14 1544 110

View Full Product Details