Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bahcc1
Entrez Gene ID
268515
Gene Name
BAH domain and coiled-coil containing 1
Gene Aliases
B930044J06, BC060615
Chromosome Location
Chr.11: 120232904 - 120292297 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198423.3 NP_940815.3 27 - 28 8111 53
GenBank mRNA AB257855.1 - 26 - 27 8113 53
AK147250.1 - 27 - 28 8111 53
AK164229.1 - 10 - 11 2352 53
BC044873.1 - 2 - 3 575 53
BC057623.1 - 2 - 3 379 53
BC060615.1 - 10 - 11 2383 53
BC072602.1 - 16 - 17 3549 53

View Full Product Details