The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139272.2 | NP_644678.2 | 15 - 16 | 1695 | 66 |
GenBank mRNA | AK046567.1 | - | 15 - 16 | 1653 | 66 |
AK145122.1 | - | 15 - 16 | 1573 | 66 | |
AK149609.1 | - | 15 - 16 | 1624 | 66 | |
AK151536.1 | - | 15 - 16 | 1486 | 66 | |
BC007172.1 | - | 14 - 15 | 1430 | 66 | |
BC053063.1 | - | 14 - 15 | 1538 | 66 | |
BC059818.1 | - | 15 - 16 | 1640 | 66 |