Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tmed6
- Entrez Gene ID
-
66269
- Gene Name
- transmembrane emp24 protein transport domain containing 6
- Gene Aliases
-
1810015P03Rik, 1810018I24Rik
- Chromosome Location
-
Chr.8: 107061484 - 107065644 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK131840.1 |
- |
|
|
|
Chr. - - [-] on Build |
69 |
View Full Product Details