The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083118.2 | NP_001076587.1 | 1 - 2 | 429 | 68 |
NM_001286200.1 | NP_001273129.1 | 1 - 2 | 429 | 68 | |
NM_009353.2 | NP_033379.2 | 1 - 2 | 429 | 68 | |
NR_104410.1 | 1 - 2 | 429 | 68 | ||
GenBank mRNA | AF003000.1 | - | 1 - 2 | 323 | 68 |
AK049507.1 | - | 1 - 2 | 429 | 68 | |
AK089419.1 | - | 1 - 2 | 403 | 69 | |
AK160421.1 | - | 1 - 2 | 74 | 68 | |
BC046284.1 | - | 1 - 2 | 418 | 68 |