The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083118.2 | NP_001076587.1 | 2 - 3 | 521 | 62 |
NM_001286200.1 | NP_001273129.1 | 2 - 3 | 521 | 62 | |
NM_009353.2 | NP_033379.2 | 2 - 3 | 521 | 62 | |
NR_104410.1 | 2 - 3 | 521 | 62 | ||
GenBank mRNA | AF003000.1 | - | 2 - 3 | 415 | 62 |
AK049507.1 | - | 2 - 3 | 521 | 62 | |
AK089419.1 | - | 2 - 3 | 496 | 62 | |
AK160421.1 | - | 2 - 3 | 166 | 62 | |
BC046284.1 | - | 2 - 3 | 510 | 62 |