Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eno4
Entrez Gene ID
226265
Gene Name
enolase 4
Gene Aliases
6430537H07Rik
Chromosome Location
Chr.19: 58939922 - 58971421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK076682.1 - 9 - 10 940 84

View Full Product Details